missing translation for 'onlineSavingsMsg'
Learn More

Acetylcholine chloride, 98+%

Catalog No. AAL0216822
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0216822 100 g
AAL0216814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0216822 Supplier Thermo Scientific Chemicals Supplier No. L0216822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 60-31-1 | C7H16ClNO2 | 181.66 g/mol

Acetylcholine chloride acts as an endogenous neurotransmitter at cholinergic synapses. The compound has been shown to have ophthalmological uses and can be broken down quickly by choliesterases. Acetylcholine influences many basic cells functions, such as mitosis, cells differentiation, cytoskeletal organization, and cell to cell contact, among other functions. It reduces mortality and plasma proinflammatory cytokines in mice with experimentally-induced sepsis.The cholinergicanti-infl ammatory mechanism is probably mediated by interaction of acetylcholine with α7n cholinoreceptor on monocytes, macrophages, and neutrophils, which decreases the levels of proinflammatory cytokines such as TNF-α, IL-1β, and IL-6.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetylcholine chloride acts as an endogenous neurotransmitter at cholinergic synapses. The compound has been shown to have ophthalmological uses and can be broken down quickly by choliesterases. Acetylcholine influences many basic cells functions, such as mitosis, cells differentiation, cytoskeletal organization, and cell to cell contact, among other functions. It reduces mortality and plasma proinflammatory cytokines in mice with experimentally-induced sepsis.The cholinergicanti-infl ammatory mechanism is probably mediated by interaction of acetylcholine with α7n cholinoreceptor on monocytes, macrophages, and neutrophils, which decreases the levels of proinflammatory cytokines such as TNF-α, IL-1β, and IL-6.

Solubility
Soluble in water (600 g/L at 20°C), alcohol, DMSO (36 mg/ml at 25°C), and ethanol (36 mg/ml at 25°C). Insoluble in ether.

Notes
Store away from moisture. Store under dry inert gas. Incompatible with heat,oxidizing agents.

Chemical Identifiers

CAS 60-31-1
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00011698
InChI Key JUGOREOARAHOCO-UHFFFAOYSA-M
Synonym acetylcholine chloride, miochol, acecoline, chloroacetylcholine, arterocoline, acecholin, ovisot, ach chloride, azetylcholinchlorid, 2-acetyloxy-n,n,n-trimethylethanaminium chloride
PubChem CID 6060
ChEBI CHEBI:2417
IUPAC Name 2-acetyloxyethyl(trimethyl)azanium;chloride
SMILES CC(=O)OCC[N+](C)(C)C.[Cl-]

Specifications

Melting Point 149°C to 152°C
Quantity 100 g
Beilstein 3571875
Sensitivity Very Hygroscopic
Merck Index 14,87
Solubility Information Soluble in water (600g/L at 20°C),alcohol,DMSO (36mg/ml at 25°C),and ethanol (36mg/ml at 25°C). Insoluble in ether.
Formula Weight 181.66
Percent Purity ≥98%
Chemical Name or Material Acetylcholine chloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-468-8
RTECSNumber FZ9800000
TSCA Yes
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only