missing translation for 'onlineSavingsMsg'
Learn More

Acetylacetonato(norbornadiene)rhodium(I)

CAS: 32354-50-0 | C12H15O2Rh | 294.16 g/mol

$197.73 - $975.55

Chemical Identifiers

CAS 32354-50-0
Molecular Formula C12H15O2Rh
Molecular Weight (g/mol) 294.16
MDL Number MFCD00075071
InChI Key IIQPRGKBFZGPIB-LWFKIUJUSA-M
IUPAC Name λ¹-rhodium(1+) bicyclo[2.2.1]hepta-2,5-diene (2Z)-4-oxopent-2-en-2-olate
SMILES [Rh+].C\C([O-])=C\C(C)=O.C1C2C=CC1C=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC382561000
SDS View Documents
Thermo Scientific Chemicals
382561000
100 mg Glass bottle
Each for $197.73
Only null left
 
AC382565000
SDS View Documents
Thermo Scientific Chemicals
382565000
500 mg Glass bottle
Each for $975.55
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

32354-50-0
294.16
IIQPRGKBFZGPIB-LWFKIUJUSA-M
[Rh+].C\C([O-])=C\C(C)=O.C1C2C=CC1C=C2
C12H15O2Rh
MFCD00075071
λ¹-rhodium(1+) bicyclo[2.2.1]hepta-2,5-diene (2Z)-4-oxopent-2-en-2-olate

Specifications

140.0°C
32354-50-0
100 mg
MFCD00075071
IIQPRGKBFZGPIB-LWFKIUJUSA-M
λ¹-rhodium(1+) bicyclo[2.2.1]hepta-2,5-diene (2Z)-4-oxopent-2-en-2-olate
294.16
Glass bottle
Yellow
Powder
C12H15O2Rh
Solubility in water: insoluble.
[Rh+].C\C([O-])=C\C(C)=O.C1C2C=CC1C=C2
294.16
Authentic
Acetylacetonato(norbornadiene)rhodium(I)
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only