missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Acetylacetonato)(norbornadiene)rhodium(I), TCI America™
$268.43 - $1336.52
Chemical Identifiers
| CAS | 32354-50-0 |
|---|---|
| Molecular Formula | C12H15O2Rh |
| Molecular Weight (g/mol) | 294.16 |
| MDL Number | MFCD00075071 |
| InChI Key | IIQPRGKBFZGPIB-LWFKIUJUSA-M |
| Synonym | (Acetylacetonato)[bicyclo[2.2.1]hepta-2,5-diene]rhodium(I) |
| IUPAC Name | λ¹-rhodium(1+) bicyclo[2.2.1]hepta-2,5-diene (2Z)-4-oxopent-2-en-2-olate |
| SMILES | [Rh+].C\C([O-])=C\C(C)=O.C1C2C=CC1C=C2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A2761200MG
|
TCI America
A2761200MG |
200 mg |
Each for $268.43
|
|
|||||
|
A27611G
|
TCI America
A27611G |
1 g |
Each for $1,336.52
|
|
|||||
Chemical Identifiers
| 32354-50-0 | |
| 294.16 | |
| IIQPRGKBFZGPIB-LWFKIUJUSA-M | |
| λ¹-rhodium(1+) bicyclo[2.2.1]hepta-2,5-diene (2Z)-4-oxopent-2-en-2-olate |
| C12H15O2Rh | |
| MFCD00075071 | |
| (Acetylacetonato)[bicyclo[2.2.1]hepta-2,5-diene]rhodium(I) | |
| [Rh+].C\C([O-])=C\C(C)=O.C1C2C=CC1C=C2 |
Specifications
| 32354-50-0 | |
| C12H15O2Rh | |
| 200 mg | |
| IIQPRGKBFZGPIB-LWFKIUJUSA-M | |
| λ¹-rhodium(1+) bicyclo[2.2.1]hepta-2,5-diene (2Z)-4-oxopent-2-en-2-olate | |
| 294.16 | |
| (Acetylacetonato)(norbornadiene)rhodium(I) |
| 175°C | |
| MFCD00075071 | |
| (Acetylacetonato)[bicyclo[2.2.1]hepta-2,5-diene]rhodium(I) | |
| [Rh+].C\C([O-])=C\C(C)=O.C1C2C=CC1C=C2 | |
| 294.16 | |
| Crystalline Powder |
Safety and Handling
TSCA : No