missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetyl Sulfide 95.0+%, TCI America™
Supplier: TCI America A097625G
Specifications
| Acetyl Sulfide | |
| Yellow | |
| C4H6O2S | |
| 25 g | |
| Diacetyl Sulfide | |
| CC(=O)SC(=O)C | |
| 118.15 | |
| 118.15 | |
| Liquid |
| 3232-39-1 | |
| 120°C | |
| MFCD00008753 | |
| 1993 | |
| DPCQJLQPDJPRCM-UHFFFAOYSA-N | |
| S-acetyl ethanethioate | |
| 76708 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 3232-39-1 | |
| 118.15 | |
| DPCQJLQPDJPRCM-UHFFFAOYSA-N | |
| 76708 | |
| CC(=O)SC(=O)C |
| C4H6O2S | |
| MFCD00008753 | |
| Diacetyl Sulfide | |
| S-acetyl ethanethioate |
Safety and Handling
TSCA : No