missing translation for 'onlineSavingsMsg'
Learn More

Acetyl-L-carnitine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7117409
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13945G 5 g
A139425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A13945G Supplier TCI America Supplier No. A13945G
Only null left

Chemical Identifiers

CAS 5080-50-2
Molecular Formula C9H19ClNO4+
Molecular Weight (g/mol) 240.704
MDL Number MFCD00082230
InChI Key JATPLOXBFFRHDN-QRPNPIFTSA-O
Synonym (R)-3-Acetoxy-4-(trimethylammonio)butyrate Hydrochloride
PubChem CID 123133819
IUPAC Name [(2S)-2-acetyloxy-3-carboxypropyl]-trimethylazanium;hydrochloride
SMILES CC(=O)OC(CC(=O)O)C[N+](C)(C)C.Cl

Specifications

Color White
Quantity 5 g
Formula Weight 239.70
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Acetyl-L-carnitine Hydrochloride