missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetyl Iodide 95.0+%, TCI America™
$484.65 - $969.30
Chemical Identifiers
| CAS | 507-02-8 |
|---|---|
| Molecular Formula | C2H3IO |
| Molecular Weight (g/mol) | 169.949 |
| MDL Number | MFCD00039399 |
| InChI Key | LEKJTGQWLAUGQA-UHFFFAOYSA-N |
| PubChem CID | 10483 |
| IUPAC Name | acetyl iodide |
| SMILES | CC(=O)I |