missing translation for 'onlineSavingsMsg'
Learn More

Acetyl Chloride 98.0+%, TCI America™

Catalog No. A0082500G
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0082500G 500 g
A0082100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0082500G Supplier TCI America Supplier No. A0082500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-36-5
Molecular Formula C2H3ClO
Molecular Weight (g/mol) 78.50
MDL Number MFCD00000719
InChI Key WETWJCDKMRHUPV-UHFFFAOYSA-N
Synonym ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662
PubChem CID 6367
ChEBI CHEBI:37580
IUPAC Name acetyl chloride
SMILES CC(Cl)=O

Specifications

Melting Point -112°C
Color Yellow
Boiling Point 52°C
Quantity 500 g
UN Number 1717
Formula Weight 78.50
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Acetyl Chloride
EINECSNumber (2)-0631
RTECSNumber AO6390000
TSCA Yes
Recommended Storage Refrigerator