missing translation for 'onlineSavingsMsg'
Learn More

Acetyl chloride, 98%

Catalog No. p-7046342
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL14013AC 25 mL
AAL14013AP 500 mL
AAL140130F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAL14013AC missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' L14013AC
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 75-36-5 | C2H3ClO | 78.50 g/mol

Acetyl chloride acts as a reagent for the preparation of esters and amides of acetic acid. It is also useful as an important reactant in Friedel-Crafts reactions as well as in the introduction of an acetyl group. It serves as a starting material in the production of pharmaceutical, new plating complexing agent, acylation agent and synthetic organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetyl chloride acts as a reagent for the preparation of esters and amides of acetic acid. It is also useful as an important reactant in Friedel-Crafts reactions as well as in the introduction of an acetyl group. It serves as a starting material in the production of pharmaceutical, new plating complexing agent, acylation agent and synthetic organic intermediates.

Solubility
Miscible with acetone, chloroform, glacial acetic acid, petroleum ether, ether and benzene.

Notes
Moisture sensitive. Incompatible with water, alcohols, oxidizing agents and strong bases.

Chemical Identifiers

CAS 75-36-5
Molecular Formula C2H3ClO
Molecular Weight (g/mol) 78.50
MDL Number MFCD00000719
InChI Key WETWJCDKMRHUPV-UHFFFAOYSA-N
Synonym ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662
PubChem CID 6367
ChEBI CHEBI:37580
IUPAC Name acetyl chloride
SMILES CC(Cl)=O

Specifications

Melting Point -112°C
Density 1.104
Boiling Point 50°C to 52°C
Flash Point 4°C (39°F)
Odor Pungent
Refractive Index 1.389
Linear Formula CH3COCl
Quantity 25 mL
UN Number UN1717
Beilstein 605303
Sensitivity Moisture sensitive
Merck Index 14,85
Solubility Information Miscible with acetone,chloroform,glacial acetic acid,petroleum ether,ether and benzene.
Formula Weight 78.5
Percent Purity 98%
Chemical Name or Material Acetyl chloride
Show More Show Less
missing translation for 'healthHazard1' H225-H314
missing translation for 'healthHazard2' GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
missing translation for 'healthHazard3' P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ACETYL CHLORIDE
EINECSNumber 200-865-6
RTECSNumber AO6390000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only