missing translation for 'onlineSavingsMsg'
Learn More

Acetyl chloride, 98%

Catalog No. p-4510206
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC151270010 1 L Glass bottle
AC151270025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC151270010 Supplier Thermo Scientific Chemicals Supplier No. 151270010
Only null left

CAS: 75-36-5 | C2H3ClO | 78.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75-36-5
Molecular Formula C2H3ClO
Molecular Weight (g/mol) 78.50
MDL Number MFCD00000719
InChI Key WETWJCDKMRHUPV-UHFFFAOYSA-N
Synonym ethanoyl chloride, acetic chloride, acetylchloride, acetic acid chloride, ch3cocl, rcra waste number u006, acetic acid, chloride, unii-qd15rno45k, ccris 4568, hsdb 662
PubChem CID 6367
ChEBI CHEBI:37580
IUPAC Name acetyl chloride
SMILES CC(Cl)=O

Specifications

Melting Point -112.0°C
Density 1.1000g/mL
Boiling Point 51.0°C
Flash Point 4°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3876 to 1.3896
Linear Formula CH3COCl
Quantity 1 L
Beilstein 02, 173
Fieser 01,11; 04,5; 05,4; 07,3; 11,4
Specific Gravity 1.1
Merck Index 15, 80
Solubility Information Solubility in water: reacts. Other solubilities: miscible with acetone, benzenz, toluene,, chloroform, ether, petroleum ether, carbon dosulfi, de and glacial acetic acid, soluble in ccl4, reacts with alcohol
Formula Weight 78.5
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Acetyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Reacts violently with water.
Corrosive to the respiratory tract.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
EINECSNumber 200-865-6

RUO – Research Use Only