Learn More
Acetyl chloride, 98%
Description
Acetyl chloride acts as a reagent for the preparation of esters and amides of acetic acid. It is also useful as an important reactant in Friedel-Crafts reactions as well as in the introduction of an acetyl group. It serves as a starting material in the production of pharmaceutical, new plating complexing agent, acylation agent and synthetic organic intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acetyl chloride acts as a reagent for the preparation of esters and amides of acetic acid. It is also useful as an important reactant in Friedel-Crafts reactions as well as in the introduction of an acetyl group. It serves as a starting material in the production of pharmaceutical, new plating complexing agent, acylation agent and synthetic organic intermediates.
Solubility
Miscible with acetone, chloroform, glacial acetic acid, petroleum ether, ether and benzene.
Notes
Moisture sensitive. Incompatible with water, alcohols, oxidizing agents and strong bases.
Specifications
Specifications
| Melting Point | -112°C |
| Density | 1.104 |
| Boiling Point | 50°C to 52°C |
| Flash Point | 4°C (39°F) |
| Odor | Pungent |
| Refractive Index | 1.389 |
| Linear Formula | CH3COCl |
| Quantity | 2500 mL |
| UN Number | UN1717 |
| Beilstein | 605303 |
| Show More |
Safety and Handling
| Hazard Category | H225-H314 |
| Hazard Statement | GHS H Statement H225-H314 Highly flammable liquid and vapor. Causes severe skin burns and eye damage. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ACETYL CHLORIDE |
| EINECSNumber | 200-865-6 |
| RTECSNumber | AO6390000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only