missing translation for 'onlineSavingsMsg'
Learn More

Acetyl Bromide 97.0+%, TCI America™

Catalog No. A008025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A008025G 25 g
A0080500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A008025G Supplier TCI America Supplier No. A008025G

Chemical Identifiers

CAS 506-96-7
Molecular Formula C2H3BrO
Molecular Weight (g/mol) 122.949
MDL Number MFCD00000114
InChI Key FXXACINHVKSMDR-UHFFFAOYSA-N
Synonym ethanoyl bromide, acetylbromide, acetic acid, bromide, ch3cobr, unii-o18v5xyo0g, hsdb 663, o18v5xyo0g, bromideacetyl, acbr, acetyl bromide
PubChem CID 10482
IUPAC Name acetyl bromide
SMILES CC(=O)Br

Specifications

Melting Point -96°C
Color Yellow
Boiling Point 77°C
Quantity 25 g
UN Number 1716
Formula Weight 122.95
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Acetyl Bromide
EINECSNumber (9)-0017
RTECSNumber AO5955000
TSCA Yes