Learn More
Acetyl bromide, 98%
CAS: 506-96-7 | C2H3BrO | 122.949 g/mol
Supplier: Thermo Scientific Chemicals A1248536
Description
Acetyl Bromide is used as an acetylating agent in the synthesis of fine chemicals, agrochemicals and pharmaceuticals. It is also used as an intermediate for dyes. Further, it reacts with alcohols and amines to produce acetate esters and acetamides. In addition to this, it is involved in the synthetic process of the anti-HIV agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAcetyl Bromide is used as an acetylating agent in the synthesis of fine chemicals, agrochemicals and pharmaceuticals. It is also used as an intermediate for dyes. Further, it reacts with alcohols and amines to produce acetate esters and acetamides. In addition to this, it is involved in the synthetic process of the anti-HIV agent.
Solubility
Miscible with ether, chloroform, benzene and acetone.
Notes
Incompatible with strong oxidizing agents, strong bases and alcohols
Specifications
| Acetyl bromide | |
| -96°C | |
| 75°C to 77°C | |
| ≥98% | |
| C2H3BrO | |
| CH3COBr | |
| 500 g | |
| 1697546 | |
| 14,82 | |
| Miscible with ether,chloroform,benzene and acetone. | |
| CC(=O)Br | |
| 122.949 | |
| 122.95 | |
| Liquid |
| 506-96-7 | |
| 1.66 | |
| >75°C (167°F) | |
| Acrid | |
| 1.45 | |
| MFCD00000114 | |
| UN1716 | |
| Moisture sensitive | |
| ethanoyl bromide, acetylbromide, acetic acid, bromide, ch3cobr, unii-o18v5xyo0g, hsdb 663, o18v5xyo0g, bromideacetyl, acbr, acetyl bromide | |
| FXXACINHVKSMDR-UHFFFAOYSA-N | |
| acetyl bromide | |
| 10482 | |
| ≥98% |
Chemical Identifiers
| 506-96-7 | |
| 122.949 | |
| FXXACINHVKSMDR-UHFFFAOYSA-N | |
| 10482 | |
| CC(=O)Br |
| C2H3BrO | |
| MFCD00000114 | |
| ethanoyl bromide, acetylbromide, acetic acid, bromide, ch3cobr, unii-o18v5xyo0g, hsdb 663, o18v5xyo0g, bromideacetyl, acbr, acetyl bromide | |
| acetyl bromide |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ACETYL BROMIDE
EINECSNumber : 208-061-7
RTECSNumber : AO5955000
TSCA : Yes
storageNote1 : Moisture Sensitive
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only