missing translation for 'onlineSavingsMsg'
Learn More

Acetoxyacetyl Chloride 95.0+%, TCI America™

Catalog No. p-7117476
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A150025G 25 g
A1500100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A150025G Supplier TCI America Supplier No. A150025G
Only null left

Chemical Identifiers

CAS 13831-31-7
Molecular Formula C4H5ClO3
Molecular Weight (g/mol) 136.531
MDL Number MFCD00011535
InChI Key HZDNNJABYXNPPV-UHFFFAOYSA-N
Synonym acetoxyacetyl chloride, 2-acetoxyacetyl chloride, unii-z4s19y2f8s, acetoxyacetylchloride, acetyl chloride, acetyloxy, acetoxyacetic acid chloride, chlorocarbonyl methyl acetate, acetoxy acetylchloride, acetyloxyacetyl chloride, pubchem20920
PubChem CID 26297
IUPAC Name (2-chloro-2-oxoethyl) acetate
SMILES CC(=O)OCC(=O)Cl

Specifications

Color Yellow
Boiling Point 160°C
Quantity 25 g
UN Number 3265
Formula Weight 136.53
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Acetoxyacetyl Chloride