missing translation for 'onlineSavingsMsg'
Learn More

Acetoxyacetyl chloride, 97%

Catalog No. AAH2579706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2579706 5 g
AAH2579714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2579706 Supplier Thermo Scientific Chemicals Supplier No. H2579706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13831-31-7 | C4H5ClO3 | 136.531 g/mol

Acetoxyacetyl chloride was used in synthesis of stabilized axial and equatorial conformers of spiro-β-lactams and glycolylhydroxamic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetoxyacetyl chloride was used in synthesis of stabilized axial and equatorial conformers of spiro-β-lactams and glycolylhydroxamic acids.

Solubility
Reacts with water violently.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, water/ moisture.

Chemical Identifiers

CAS 13831-31-7
Molecular Formula C4H5ClO3
Molecular Weight (g/mol) 136.531
MDL Number MFCD00011535
InChI Key HZDNNJABYXNPPV-UHFFFAOYSA-N
Synonym acetoxyacetyl chloride, 2-acetoxyacetyl chloride, unii-z4s19y2f8s, acetoxyacetylchloride, acetyl chloride, acetyloxy, acetoxyacetic acid chloride, chlorocarbonyl methyl acetate, acetoxy acetylchloride, acetyloxyacetyl chloride, pubchem20920
PubChem CID 26297
IUPAC Name (2-chloro-2-oxoethyl) acetate
SMILES CC(=O)OCC(=O)Cl

Specifications

Density 1.27
Boiling Point 55°C (12 mmHg)
Flash Point 71°C (160°F)
Refractive Index 1.4271
Linear Formula CH3CO2CH2COCl
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Reacts with water violently.
Formula Weight 136.53
Percent Purity 97%
Chemical Name or Material Acetoxyacetyl chloride
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H335-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 237-542-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only