missing translation for 'onlineSavingsMsg'
Learn More

Acetoxyacetone, 97%

Catalog No. AAH31346AD
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH31346AD 50 mL
AAH31346AB 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH31346AD Supplier Thermo Scientific Chemicals Supplier No. H31346AD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 592-20-1 | C5H8O3 | 116.116 g/mol

Acetoxyacetone is an building block in the synthesis of various pharmaceutical and organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetoxyacetone is an building block in the synthesis of various pharmaceutical and organic compounds.

Solubility
Soluble in diethyl ether, ethyl acetate and dichloromethane.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 592-20-1
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD00042848
InChI Key DBERHVIZRVGDFO-UHFFFAOYSA-N
Synonym acetoxyacetone, acetonyl acetate, acetol acetate, 2-propanone, 1-acetyloxy, acetoxypropanone, o-acetylacetol, 1-acetoxyacetone, acetylmethyl acetate, acetoxy-2-propanone, 1-acetoxy-2-propanone
PubChem CID 11593
IUPAC Name 2-oxopropyl acetate
SMILES CC(=O)COC(=O)C

Specifications

Density 1.075
Boiling Point 174°C to 176°C
Flash Point 71°C (160°F)
Refractive Index 1.417
Quantity 50 mL
Solubility Information Soluble in diethyl ether,ethyl acetate and dichloromethane.
Formula Weight 116.12
Percent Purity 97%
Chemical Name or Material Acetoxyacetone
Hazard Category H227-H302+H332-H315-H319
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 209-746-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only