missing translation for 'onlineSavingsMsg'
Learn More

Acetoxy-2-propanone (stabilized with Mg6Al2(OH)16CO3.4H2O) 97.0+%, TCI America™

Catalog No. a106625ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
Catalog No. Quantity
A106625ML 25 mL
1 options

Catalog No. A106625ML

Supplier: TCI America A106625ML

Only null left

Chemical Identifiers

CAS 592-20-1
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD00042848
InChI Key DBERHVIZRVGDFO-UHFFFAOYSA-N
Synonym acetoxyacetone, acetonyl acetate, acetol acetate, 2-propanone, 1-acetyloxy, acetoxypropanone, o-acetylacetol, 1-acetoxyacetone, acetylmethyl acetate, acetoxy-2-propanone, 1-acetoxy-2-propanone
PubChem CID 11593
IUPAC Name 2-oxopropyl acetate
SMILES CC(=O)COC(=O)C

Specifications

Color Yellow
Boiling Point 174°C
Quantity 25 mL
Formula Weight 116.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Acetoxy-2-propanone (stabilized with Mg6Al2(OH)16CO3.4H2O)