missing translation for 'onlineSavingsMsg'
Learn More

Acetoxime O-(2,4,6-Trimethylphenylsulfonate) 98.0+%, TCI America™

Catalog No. p-7117438
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14415G 5 g
A144125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14415G Supplier TCI America Supplier No. A14415G

Chemical Identifiers

CAS 81549-07-7
Molecular Formula C12H17NO3S
Molecular Weight (g/mol) 255.332
MDL Number MFCD01321127
InChI Key BCPWUOSBRCQZFB-UHFFFAOYSA-N
Synonym Acetone O-(2,4,6-Trimethylphenylsulfonyl)oxime, O-(2,4,6-Trimethylphenylsulfonyl)acetoxime
PubChem CID 279802
IUPAC Name (propan-2-ylideneamino) 2,4,6-trimethylbenzenesulfonate
SMILES CC1=CC(=C(C(=C1)C)S(=O)(=O)ON=C(C)C)C

Specifications

Melting Point 96°C
Color White-Yellow
Quantity 5 g
Formula Weight 255.33
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Acetoxime O-(2,4,6-Trimethylphenylsulfonate)