missing translation for 'onlineSavingsMsg'
Learn More

Acetovanillone, 98%

Catalog No. p-3777129
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC102420250 25 g Glass bottle
AC102421000 100 g Glass bottle
AC102425000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC102420250 Supplier Thermo Scientific Chemicals Supplier No. 102420250

CAS: 498-02-2 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 498-02-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00008747
InChI Key DFYRUELUNQRZTB-UHFFFAOYSA-N
Synonym acetovanillone, apocynin, 4'-hydroxy-3'-methoxyacetophenone, 1-4-hydroxy-3-methoxyphenyl ethanone, acetoguaiacone, acetoguaiacon, apocynine, 4-acetyl-2-methoxyphenol, acetovanilone, acetovanyllon
PubChem CID 2214
ChEBI CHEBI:2781
IUPAC Name 1-(4-hydroxy-3-methoxyphenyl)ethanone
SMILES COC1=CC(=CC=C1O)C(C)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 112.0°C to 116.0°C
Color Yellow to Brown
Boiling Point 295.0°C to 300.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HOC6H3(OCH3)COCH3
Quantity 25 g
Beilstein 08, 272
Merck Index 15, 728
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in ethanol,acetone,benzene,very soluble in diethyl ether,chloroform,practically insoluble in petroleum ether
Formula Weight 166.18
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Acetovanillone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 207-854-5

RUO – Research Use Only