missing translation for 'onlineSavingsMsg'
Learn More

Acetophenone 98.5+%, TCI America™

Catalog No. p-7116689
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A006125G 25 g
A0061500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A006125G Supplier TCI America Supplier No. A006125G
Only null left

Chemical Identifiers

CAS 98-86-2
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00008724
InChI Key KWOLFJPFCHCOCG-UHFFFAOYSA-N
Synonym acetophenone, methyl phenyl ketone, acetylbenzene, phenyl methyl ketone, ethanone, 1-phenyl, hypnone, benzoyl methide, acetophenon, 1-phenylethan-1-one, acetylbenzol
PubChem CID 7410
ChEBI CHEBI:27632
IUPAC Name 1-phenylethanone
SMILES CC(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 202°C
Quantity 25 g
Formula Weight 120.15
Percent Purity ≥98.5% (GC)
Physical Form Liquid
Chemical Name or Material Acetophenone
EINECSNumber (3)-1231
RTECSNumber AM5250000
TSCA Yes