Learn More
Acetophenone, 99%
CAS: 98-86-2 | C8H8O | 120.151 g/mol
Supplier: Thermo Scientific Chemicals A127270D
Description
Acetophenone is used as a raw material in the synthesis of active pharmaceutical ingredients such as dextropropoxyphene and phenylpropanolamine. It is employed in organic synthesis and as a polymerization catalyst for olefins. It is involved in the preparation of styrene, 1-phenylethanol. It is also used as a active component of coatings and inks. It reacts with formaldehyde to prepare resins. Further, it is used in textile coatings, in perfumes for orange blossom odor. In addition to this, it is used as a solvent in plastics, adhesives and a flavoring agent in foods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAcetophenone is used as a raw material in the synthesis of active pharmaceutical ingredients such as dextropropoxyphene and phenylpropanolamine. It is employed in organic synthesis and as a polymerization catalyst for olefins. It is involved in the preparation of styrene, 1-phenylethanol. It is also used as a active component of coatings and inks. It reacts with formaldehyde to prepare resins. Further, it is used in textile coatings, in perfumes for orange blossom odor. In addition to this, it is used as a solvent in plastics, adhesives and a flavoring agent in foods.
Solubility
Miscible with water, chloroform, ethanol, ether, benzene and acetone. Slightly miscible with concentrated sulfuric acid.
Notes
Light sensitive. Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
Specifications
| Acetophenone | |
| 19°C to 20°C | |
| 201°C to 203°C | |
| C8H8O | |
| C6H5COCH3 | |
| 10,000 mL | |
| acetophenone, methyl phenyl ketone, acetylbenzene, phenyl methyl ketone, ethanone, 1-phenyl, hypnone, benzoyl methide, acetophenon, 1-phenylethan-1-one, acetylbenzol | |
| KWOLFJPFCHCOCG-UHFFFAOYSA-N | |
| 1-phenylethanone | |
| 7410 | |
| 120.15 |
| 98-86-2 | |
| 1.03 | |
| 76°C (169°F) | |
| 1.5325 | |
| MFCD00008724 | |
| 14,73 | |
| Miscible with water,chloroform,ethanol,ether,benzene and acetone. Slightly miscible with concentrated sulfuric acid. | |
| CC(=O)C1=CC=CC=C1 | |
| 120.151 | |
| CHEBI:27632 | |
| 99% |
Chemical Identifiers
| 98-86-2 | |
| 120.151 | |
| KWOLFJPFCHCOCG-UHFFFAOYSA-N | |
| 7410 | |
| 1-phenylethanone |
| C8H8O | |
| MFCD00008724 | |
| acetophenone, methyl phenyl ketone, acetylbenzene, phenyl methyl ketone, ethanone, 1-phenyl, hypnone, benzoyl methide, acetophenon, 1-phenylethan-1-one, acetylbenzol | |
| CHEBI:27632 | |
| CC(=O)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H302-H319-H227
Harmful if swallowed.
Causes serious eye irritation.
Combustible liquid.
P210-P264b-P270-P280-P301+P312-P305+P351+P338-P330-P370+P378q-P501c
H227-H302-H319
EINECSNumber : 202-708-7
RTECSNumber : AM5250000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only