Learn More
Acetophenone, 98%, pure
CAS: 98-86-2 | C8H8O | 120.15 g/mol
Supplier: Thermo Scientific Chemicals 102412500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Acetophenone | |
| 97.5 | |
| 19.6°C | |
| 1.0280g/mL | |
| 77°C | |
| 97.5% min. (GC) | |
| C8H8O | |
| C6H5COCH3 | |
| 250 mL | |
| 04,5 | |
| 15, 71 | |
| Solubility in water: 5.5g/L (20°C). Other solubilities: soluble in conc. h2so4,freely soluble in alcohol,chloroform,ether and,glycerol | |
| CC(=O)C1=CC=CC=C1 | |
| 120.15 | |
| 1.8 mPa.s (20°C) | |
| 120.15 | |
| Pure |
| 98-86-2 | |
| 100.0 | |
| Colorless to Yellow | |
| 202.0°C | |
| Authentic | |
| Glass bottle | |
| 1.5315 to 1.5340 | |
| MFCD00008724 | |
| 07, IV, 619 | |
| 1.028 | |
| acetophenone, methyl phenyl ketone, acetylbenzene, phenyl methyl ketone, ethanone, 1-phenyl, hypnone, benzoyl methide, acetophenon, 1-phenylethan-1-one, acetylbenzol | |
| KWOLFJPFCHCOCG-UHFFFAOYSA-N | |
| 1-phenylethanone | |
| 7410 | |
| CHEBI:27632 | |
| 98% | |
| Liquid |
Chemical Identifiers
| 98-86-2 | |
| KWOLFJPFCHCOCG-UHFFFAOYSA-N | |
| 7410 | |
| 1-phenylethanone |
| MFCD00008724 | |
| acetophenone, methyl phenyl ketone, acetylbenzene, phenyl methyl ketone, ethanone, 1-phenyl, hypnone, benzoyl methide, acetophenon, 1-phenylethan-1-one, acetylbenzol | |
| CHEBI:27632 | |
| CC(=O)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
EINECSNumber : 202-708-7
RUO – Research Use Only