missing translation for 'onlineSavingsMsg'
Learn More

Acetonyltriphenylphosphonium Chloride 98.0+%, TCI America™

Catalog No. A130525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A130525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A130525G Supplier TCI America Supplier No. A130525G

Chemical Identifiers

CAS 1235-21-8
Molecular Formula C21H20ClOP
Molecular Weight (g/mol) 354.81
MDL Number MFCD00011813
InChI Key XAMZZEBAJZJERT-UHFFFAOYSA-M
Synonym acetonyltriphenylphosphonium chloride, 2-oxopropyl triphenylphosphonium chloride, 2-oxopropyltriphenylphosphonium chloride, phosphonium, acetonyltriphenyl-, chloride, acetonyl triphenylphosphonium chloride, phosphonium, 2-oxopropyl triphenyl-, chloride, acetonyltriphenylphosphoniumchloride, 2-oxopropyl triphenylphosphanium chloride, 1-triphenylphosphino acetone, chloride, acmc-1c01z
PubChem CID 196994
IUPAC Name (2-oxopropyl)triphenylphosphanium chloride
SMILES [Cl-].CC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 249°C
Quantity 25 g
Formula Weight 354.81
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Acetonyltriphenylphosphonium Chloride
RTECSNumber TA1841000
TSCA No