missing translation for 'onlineSavingsMsg'
Learn More

Acetonyltriphenylphosphonium chloride, 99%

CAS: 1235-21-8 | C21H20ClOP | 354.81 g/mol

Supplier:  Thermo Scientific Chemicals A1469122

 View more versions of this product

Catalog No. AAA1469122


Only null left
Explore available promotions

Description

Description

Acetonyltriphenylphosphonium chloride is used to investigate the antimycobacterial activities of Me alkenyl quinolones. It is used as a reactant for synthesis of unsaturated esters and ketones containing multiple aromatic and heteroaromatic rings using Suzuki coupling - Wittig olefination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetonyltriphenylphosphonium chloride is used to investigate the antimycobacterial activities of Me alkenyl quinolones. It is used as a reactant for synthesis of unsaturated esters and ketones containing multiple aromatic and heteroaromatic rings using Suzuki coupling - Wittig olefination.

Solubility
Soluble in hot water.

Notes
It is hygroscopic. Incompatible with oxidizing agents.
Specifications

Specifications

Acetonyltriphenylphosphonium chloride
244°C to 248°C
MFCD00011813
Hygroscopic
Soluble in hot water.
[Cl-].CC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
354.81
354.82
1235-21-8
C21H20ClOP
100 g
acetonyltriphenylphosphonium chloride, 2-oxopropyl triphenylphosphonium chloride, 2-oxopropyltriphenylphosphonium chloride, phosphonium, acetonyltriphenyl-, chloride, acetonyl triphenylphosphonium chloride, phosphonium, 2-oxopropyl triphenyl-, chloride, acetonyltriphenylphosphoniumchloride, 2-oxopropyl triphenylphosphanium chloride, 1-triphenylphosphino acetone, chloride, acmc-1c01z
XAMZZEBAJZJERT-UHFFFAOYSA-M
2-oxopropyl(triphenyl)phosphanium;chloride
196994
99%

Chemical Identifiers

1235-21-8
354.81
XAMZZEBAJZJERT-UHFFFAOYSA-M
196994
[Cl-].CC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C21H20ClOP
MFCD00011813
acetonyltriphenylphosphonium chloride, 2-oxopropyl triphenylphosphonium chloride, 2-oxopropyltriphenylphosphonium chloride, phosphonium, acetonyltriphenyl-, chloride, acetonyl triphenylphosphonium chloride, phosphonium, 2-oxopropyl triphenyl-, chloride, acetonyltriphenylphosphoniumchloride, 2-oxopropyl triphenylphosphanium chloride, 1-triphenylphosphino acetone, chloride, acmc-1c01z
2-oxopropyl(triphenyl)phosphanium;chloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 214-974-1

RTECSNumber : TA1841000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only