missing translation for 'onlineSavingsMsg'
Learn More

Acetonyltriphenylphosphonium chloride, 99%

Catalog No. AAA1469136
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1469136 500 g
AAA1469114 25 g
AAA1469122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1469136 Supplier Thermo Scientific Chemicals Supplier No. A1469136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1235-21-8 | C21H20ClOP | 354.81 g/mol

Acetonyltriphenylphosphonium chloride is used to investigate the antimycobacterial activities of Me alkenyl quinolones. It is used as a reactant for synthesis of unsaturated esters and ketones containing multiple aromatic and heteroaromatic rings using Suzuki coupling - Wittig olefination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetonyltriphenylphosphonium chloride is used to investigate the antimycobacterial activities of Me alkenyl quinolones. It is used as a reactant for synthesis of unsaturated esters and ketones containing multiple aromatic and heteroaromatic rings using Suzuki coupling - Wittig olefination.

Solubility
Soluble in hot water.

Notes
It is hygroscopic. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1235-21-8
Molecular Formula C21H20ClOP
Molecular Weight (g/mol) 354.81
MDL Number MFCD00011813
InChI Key XAMZZEBAJZJERT-UHFFFAOYSA-M
Synonym acetonyltriphenylphosphonium chloride, 2-oxopropyl triphenylphosphonium chloride, 2-oxopropyltriphenylphosphonium chloride, phosphonium, acetonyltriphenyl-, chloride, acetonyl triphenylphosphonium chloride, phosphonium, 2-oxopropyl triphenyl-, chloride, acetonyltriphenylphosphoniumchloride, 2-oxopropyl triphenylphosphanium chloride, 1-triphenylphosphino acetone, chloride, acmc-1c01z
PubChem CID 196994
IUPAC Name 2-oxopropyl(triphenyl)phosphanium;chloride
SMILES [Cl-].CC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 244°C to 248°C
Quantity 500 g
Sensitivity Hygroscopic
Solubility Information Soluble in hot water.
Formula Weight 354.82
Percent Purity 99%
Chemical Name or Material Acetonyltriphenylphosphonium chloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-974-1
RTECSNumber TA1841000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only