missing translation for 'onlineSavingsMsg'
Learn More

Acetonylacetone 97.0+%, TCI America™

Catalog No. p-7130340
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H010125ML 25 mL
H0101100ML 100 mL
H0101500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H010125ML Supplier TCI America Supplier No. H010125ML
Only null left

Chemical Identifiers

CAS 110-13-4
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00008792
InChI Key OJVAMHKKJGICOG-UHFFFAOYSA-N
Synonym 2,5-hexanedione, acetonylacetone, 1,2-diacetylethane, acetonyl acetone, diacetonyl, 2,5-diketohexane, 2,5-hexadione, acetone, acetonyl, 2,5-hexandione, alpha,beta-diacetylethane
PubChem CID 8035
ChEBI CHEBI:85014
IUPAC Name hexane-2,5-dione
SMILES CC(=O)CCC(=O)C

Specifications

Melting Point -9°C
Boiling Point 188°C
Quantity 25 mL
Formula Weight 114.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Acetonylacetone
EINECSNumber (2)-0563
RTECSNumber MO3150000
TSCA Yes