missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonylacetone 97.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | -9°C |
| Boiling Point | 188°C |
| Quantity | 25 mL |
| Formula Weight | 114.14 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Acetonylacetone |
Sécurité et manipulation
| EINECSNumber | (2)-0563 |
| RTECSNumber | MO3150000 |
| TSCA | Yes |