Learn More
Acetonylacetone, 97%
CAS: 110-13-4 | C6H10O2 | 114.14 g/mol
Supplier: Thermo Scientific Chemicals 102401000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Acetonylacetone | |
| 110-13-4 | |
| 100.0 | |
| Yellow to Brown | |
| 185.0°C to 193.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4250 to 1.4270 | |
| MFCD00008792 | |
| 01,788 | |
| 15,7 | |
| Solubility in water: miscible. Other solubilities: miscible with alcohol, ether, ethanol and, diethyl ether, miscible in organic solvents | |
| CC(=O)CCC(=O)C | |
| 114.14 | |
| 1.43 mPa.s (25°C) | |
| 114.14 | |
| Liquid |
| 97% | |
| 96.0 | |
| -6.0°C | |
| 0.9730g/mL | |
| 78°C | |
| 97% | |
| C6H10O2 | |
| CH3COCH2CH2COCH3 | |
| 100 mL | |
| 0.973 | |
| 2,5-hexanedione, acetonylacetone, 1,2-diacetylethane, acetonyl acetone, diacetonyl, 2,5-diketohexane, 2,5-hexadione, acetone, acetonyl, 2,5-hexandione, alpha,beta-diacetylethane | |
| OJVAMHKKJGICOG-UHFFFAOYSA-N | |
| hexane-2,5-dione | |
| 8035 | |
| CHEBI:85014 | |
| 97% |
Chemical Identifiers
| 110-13-4 | |
| 114.14 | |
| OJVAMHKKJGICOG-UHFFFAOYSA-N | |
| 8035 | |
| hexane-2,5-dione |
| C6H10O2 | |
| MFCD00008792 | |
| 2,5-hexanedione, acetonylacetone, 1,2-diacetylethane, acetonyl acetone, diacetonyl, 2,5-diketohexane, 2,5-hexadione, acetone, acetonyl, 2,5-hexandione, alpha,beta-diacetylethane | |
| CHEBI:85014 | |
| CC(=O)CCC(=O)C |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clot
GHS Signal Word: Warning
EINECSNumber : 203-738-3
RTECSNumber : MO3150000
TSCA : TSCA
RUO – Research Use Only