missing translation for 'onlineSavingsMsg'
Learn More

Acetonylacetone, 97%

Catalog No. AC102400050
Change view
Click to view available options
Quantity:
5 mL
100 mL
250 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC102400050 5 mL Glass bottle
AC102401000 100 mL Glass bottle
AC102402500 250 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC102400050 Supplier Thermo Scientific Chemicals Supplier No. 102400050
Only null left

CAS: 110-13-4 | C6H10O2 | 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-13-4
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00008792
InChI Key OJVAMHKKJGICOG-UHFFFAOYSA-N
Synonym 2,5-hexanedione, acetonylacetone, 1,2-diacetylethane, acetonyl acetone, diacetonyl, 2,5-diketohexane, 2,5-hexadione, acetone, acetonyl, 2,5-hexandione, alpha,beta-diacetylethane
PubChem CID 8035
ChEBI CHEBI:85014
IUPAC Name hexane-2,5-dione
SMILES CC(=O)CCC(=O)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -6.0°C
Color Yellow to Brown
Density 0.9730g/mL
Boiling Point 185.0°C to 193.0°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4250 to 1.4270
Linear Formula CH3COCH2CH2COCH3
Quantity 5 mL
Beilstein 01,788
Merck Index 15,7
Specific Gravity 0.973
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol, ether, ethanol and, diethyl ether, miscible in organic solvents
Viscosity 1.43 mPa.s (25°C)
Formula Weight 114.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Acetonylacetone, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clot
EINECSNumber 203-738-3
RTECSNumber MO3150000
TSCA TSCA

RUO – Research Use Only