missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, UHPLC-MS
Unique signal-to-noise specification for the chemical industry where the solvent quality is linked directly to the sensitivity of the mass spectrometer. | CAS: 75-05-8 | C2H3N | 41.053 g/mol
Supplier: Thermo Fisher Scientific A9561
Description
Acetonitrile, C2H3N, CAS Number-75-05-8, cyanure de methyl, methanecarbonitrile, cyanomethane, acetonitrile, mecn, methane, cyano, methyl cyanide, ethanenitrile, methylkyanid, ethyl nitrile, 1L
Specifications
| Acetonitrile | |
| 75-05-8 | |
| Colorless | |
| 81°C | |
| −40°C (-40°F) | |
| Liquid | |
| 99.9% minimum by GC | |
| C2H3N | |
| 97mbar at 20°C | |
| 0.01% max. | |
| WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
| acetonitrile | |
| 6342 | |
| UHPLC-MS |
| -46°C | |
| ≤0.005 AU at 235nm, ≤0.15 AU at 195nm, ≤1.00 AU at 190nm, ≤0.005 AU at 254nm, ≤0.015 AU at 220nm, ≤0.015 AU at 225nm, ≤0.015 AU at 220nm, ≤0.03 AU at 210nm, ≤0.025 AU at 215nm, ≤0.03 AU at 210nm, ≤0.04 AU at 205nm, ≤0.05 AU at 200nm | |
| 0.781 g/cm3 | |
| 1 L | |
| Clear Borosilicate Glass Bottle | |
| 0.8ppm max. | |
| 0.008meq/g max. | |
| 0.0006meq/g max. | |
| 0.36 cP at 20°C | |
| methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
| CC#N | |
| 41.053 | |
| CHEBI:38472 |
RUO – Research Use Only