missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile/TFA 0.1% Mobile Phase, MilliporeSigma™

Catalog No. MTX1277P1
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MTX1277P1 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MTX1277P1 Supplier Technidata Supplier No. US1TX1277P/1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

Molecular Formula CF3COOH in CH3CN

Specifications

Quantity 4 L
Physical Form Liquid
Linear Formula CFCH3COOH in CHCH3CN
Formula Weight 114.2
Chemical Name or Material Acetonitrile/TFA 0.1% Mobile Phase, Mobile Phase
Recommended Storage Room Temperature