missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99.5% (GC), Honeywell Riedel-de Haën™
Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99.5% (GC)
Supplier: Honeywell-Riedel-de Haen 330194X25L
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
| Acetonitrile | |
| Reag. Ph. Eur. | |
| Free acid (as CH3COOH): ≤0.001% | |
| 0.786g/mL at 25°C | |
| 4 x 2.5 L | |
| 2°C (35.6°F) | |
| Glass Bottle | |
| C2H3N | |
| MFCD00001878 | |
| UN1648 | |
| methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
| CC#N | |
| 41.053 | |
| CHEBI:38472 | |
| ≥99.5% (GC) |
| -48°C | |
| 75-05-8 | |
| Free alkali (as NH3): ≤0.001% | |
| 82°C | |
| 0.16 | |
| 973°F | |
| n20/D 1.3435 to 1.3445 | |
| CH3CN | |
| 72.8mmHg (20°C) | |
| 741857 | |
| WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
| acetonitrile | |
| 6342 | |
| 41.05g/mol | |
| ACS/puriss. p.a. |
Chemical Identifiers
| 75-05-8 | |
| 41.053 | |
| WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
| 6342 | |
| acetonitrile |
| C2H3N | |
| MFCD00001878 | |
| methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
| CHEBI:38472 | |
| CC#N |
Safety and Handling
P210-P280-P305 + P351 + P338
H225-H302 + H312 + H332-H319
Danger
DOTInformation : 3/PGII
EINECSNumber : 200-835-2
RTECSNumber : AL7700000