missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.8%, Solstice

Catalog No. 6000287
Change view
Click to view available options
Quantity:
19 L
Packaging:
Metal Can
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000287 19 L Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6000287 Supplier Solstice Supplier No. PP016TC19

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

CAS Max % 100.0000%
Density 0.7822 g/cm3
Quantity 19 L
Flash Point 6°C
Packaging Metal Can
UN Number UN1648
Beilstein 741857
Boiling Point 82°C
Formula Weight 41
Percent Purity >99.8%
Grade ACS
Chemical Name or Material Acetonitrile, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.8%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile