missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, OmniSolv™, Non−UV, MilliporeSigma™

Catalog No. 50947943
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle (Pre-packed)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
50947943 4 L Glass Bottle (Pre-packed)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50947943 Supplier Technidata Supplier No. AX01551

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -45.7°C
Quantity 4 L
Flash Point 2°C
Packaging Glass Bottle (Pre-packed)
Physical Form Clear liquid
Titratable Acid 8.3μeq/g (max)
Vapor Pressure 97 hPa (20°C)
Water 0.10% max
Boiling Point 81.6°C (1013 hPa)
Formula Weight 41.05
Percent Purity 99.9% min
Chemical Name or Material Acetonitrile, Non UV
Show More Show Less
EINECSNumber 200-835-2
RTECSNumber AL7700000