missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, LC-MS Ultra CHROMASOLV™, Honeywell Riedel-de Haën™

Catalog No. p-7071821
Change view
Click to view available options
Quantity:
1 L
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000265 1 L
6000266 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000265 Supplier Honeywell Chemicals Supplier No. 142611L
Only null left

CHROMASOLV™ LC-MS Ultra, tested for UHPLC-MS

  • For the most demanding LC-MS and UHPLC applications
  • Tested for UHPLC-MS suitability
  • Highest solvent purity for low detection limits
  • Extremely low MS baseline noise
  • Exemplary lot-to-lot reproducibility
  • Low levels of trace metals
  • Packaged in clear borosilicate glass bottles

 

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Density 0.786g/mL at 25°C
Quantity 1 L
Explosion Limit 0.16
Flash Point 2°C (35.6°F)
Ignition Point 973°F
Linear Formula CH3CN
Vapor Pressure 72.8mmHg (20°C)
UN Number 1648
Beilstein 741857
Solubility Information Completely souble in water
Formula Weight 41.05g/mol
Grade LC-MS Ultra CHROMASOLV
Chemical Name or Material Acetonitrile
Show More Show Less
Hazard Statement P210-P280-P305 + P351 + P338
Precautionary Statement H225-H302 + H312 + H332-H319
EINECSNumber 200-835-2
RTECSNumber AL7700000