missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, LC-MS Grade For Liquid Chromatography- Mass Spectrometry, OmniSolv™, MilliporeSigma™

Catalog No. 361041849
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
361041849 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 361041849 Supplier Technidata Supplier No. AX01566
Only null left

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -45.7°C
Boiling Point 81.6°C (1013 hPa)
Quantity 1 L
Flash Point 2°C
Packaging Glass Bottle
Physical Form Clear Liquid
Residue after Evaporation 1ppm
Titratable Acid 1µeq/g
Vapor Pressure 97 hPa (20°C)
Water 0.02%
Formula Weight 41.05
Percent Purity <99.9%
Chemical Name or Material Acetonitrile, LC-MS
Show More Show Less
EINECSNumber 200-835-2
RTECSNumber AL7700000