Learn More
Acetonitrile, HPLC Grade (Far UV), 99.8+%
Suitable For Far UV Analysis | CAS: 75-05-8 | C2H3N | 41.053 g/mol
$206.53 - $616.10
Chemical Identifiers
| CAS | 75-05-8 |
|---|---|
| Molecular Formula | C2H3N |
| Molecular Weight (g/mol) | 41.053 |
| MDL Number | MFCD00001878 |
| InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
| Synonym | methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn |
| PubChem CID | 6342 |
| ChEBI | CHEBI:38472 |
| IUPAC Name | acetonitrile |
| SMILES | CC#N |
Description
It is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 75-05-8 | |
| 41.053 | |
| WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
| 6342 | |
| acetonitrile |
| C2H3N | |
| MFCD00001878 | |
| methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
| CHEBI:38472 | |
| CC#N |
Specifications
| °C to 48°C to°C to 45°C | |
| Colorless | |
| 80°C to 82°C | |
| 5°C (41°F) | |
| Pungent | |
| 1.344 | |
| C2H3N | |
| 73mm Hg at 20°C | |
| 741857 | |
| Miscible with water,methanol,methyl acetate,ethyl acetate and chloroform. Immiscible with saturated hydrocarbons. | |
| CC#N | |
| 41.053 | |
| CHEBI:38472 | |
| HPLC |
| 75-05-8 | |
| 0.786 | |
| 1 L | |
| 525°C (977°F) | |
| Liquid | |
| >99.8% | |
| MFCD00001878 | |
| 1648; Colorless | |
| methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
| WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
| acetonitrile | |
| 6342 | |
| 41.05 | |
| Acetonitrile |
Safety and Handling
P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P330-P363-P370+P378q-P501c
H225-H302+H312+H332-H319
DOTInformation : DOT Class: 3, Packaging Group: II
EINECSNumber : 200-835-2
RTECSNumber : AL7700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only