missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, HPLC Grade, ≥ 99.8%, LabChem™

Catalog No. LC104602
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC104602 1 L Amber Glass
LC104604 4 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC104602 Supplier LabChem Supplier No. LC104602
Only null left

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Absorbance Passes Test
Color Colorless
CAS Max % 100
Acidity ≤8μeq/g
Alkalinity ≤0.6μeq/g
Decomposition Information Carbon dioxide Carbon monoxide Nitrogen oxides cyanides
Density 787kg/m3
Quantity 1 L
Identification Passes Test
Packaging Amber Glass
Physical Form Liquid
Residue after Evaporation ≤5ppm
Specific Gravity 0.79g/mL
Linear Formula CH3CN
Viscosity 3.80mm2/s
UN Number UN1648
Water ≤0.1%
Solubility Information Soluble in water
Formula Weight 41.05
Percent Purity ≥99.8%
Grade HPLC
Chemical Name or Material Acetonitrile
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Toxic in contact with skin.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/ hot surfaces.
No smoking.
Ground/Bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Wear protective gloves, protective clothing, eye protection, face protection.
Store locked up in a well-ventilated place.
Keep cool.
Keep container tightly closed.
If on skin (or hair): Take off immediately all contaminated clothing.
Wash skin with plenty of soap and water.
Call a poison center/doctor if you feel unwell.
Wash contaminated clothing before reuse.
In case of fire: Use carbon dioxide, powder, alcohol-resistant foam to extinguish.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature