missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, Gradient grade, 99.9% min, OmniSolv, MilliporeSigma™

Catalog No. MAX01426
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Glass Bottle
NOWPak™
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAX01426 1 L Glass Bottle
MAX01421 4 L Glass Bottle
MAX0142NP20 20 L NOWPak™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MAX01426 Supplier Technidata Supplier No. AX01426

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Color Colorless
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range 99.9% min.
Formula Weight 41.05
Chemical Name or Material Acetonitrile