missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, Gradient grade, 99.9% min, OmniSolv, MilliporeSigma™
Specifications
Specifications
| Color | Colorless |
| Quantity | 4 L |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Assay Percent Range | 99.9% min. |
| Formula Weight | 41.05 |
| Chemical Name or Material | Acetonitrile |