missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, For Spectroscopy Uvasol™, MilliporeSigma™
Specifications
Specifications
| Melting Point | -45.7°C |
| Color | Colorless |
| Density | 0.79g/cm3 at 20°C |
| Quantity | 2.5 L |
| Ignition Point | 524°C (975°F) |
| Odor | Ether-like |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Vapor Pressure | 97 hPa (20°C) |
| UN Number | 1648 |
| Show More |
Safety and Handling
| Hazard Category | H225: Highly flammable liquid and vapour. |
| Hazard Statement | H302 + H312 + H332: Harmful if swallowed, in contact with skin or if inhaled |
| Precautionary Statement | H319: Causes serious eye irritation. |
| Recommended Storage | 15° to 25°C (59° to 77°F); Keep container tightly closed. Keep container in a cool, well-ventilated place. Keep away from heat and source of ignition. Protected from light. |