missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile 99.5+%, TCI America™

Catalog No. A0293100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0293100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A0293100ML Supplier TCI America Supplier No. A0293100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46°C
Color Colorless
Boiling Point 82°C
Quantity 100 mL
UN Number 1648
Formula Weight 41.05
Percent Purity ≥99.5% (GC)
Physical Form Liquid
Chemical Name or Material Acetonitrile [for Spectrophotometry]
EINECSNumber (2)-1508
RTECSNumber AL7700000
TSCA Yes