missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, For gas chromatography ECD and FID SupraSolv™, MilliporeSigma™

Catalog No. M1000171000
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1000171000 1 L Glass Bottle
M1000172500 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1000171000 Supplier Technidata Supplier No. 1.00017.1000

For gas chromatography ECD and FID SupraSolv™

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -45.7°C
Color Colorless
Density 0.79g/cm3 at 20°C
Quantity 1 L
Flash Point 2°C (36°F)
Ignition Point 524°C (975°F)
Odor Ether-like
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.34
Vapor Pressure 97 hPa (20°C)
UN Number 1648
Boiling Point 81.6°C
Solubility Information Soluble in water at 20°C
Formula Weight 41.05
Chemical Name or Material Acetonitrile
Show More Show Less
Hazard Category H225: Highly flammable liquid and vapour.
Hazard Statement H302 + H312 + H332: Harmful if swallowed, in contact with skin or if inhaled
Precautionary Statement H319: Causes serious eye irritation.
Recommended Storage 15° to 25°C (59° to 77°F); Keep container tightly closed. Keep container in a cool, well-ventilated place. Keep away from heat and source of ignition. Protected from light.