missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, Exceeds A.C.S. Specifications, HPLC Grade, 99.9%, Spectrum™ Chemical

Catalog No. 18608409
Change view
Click to view available options
Quantity:
4 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608409 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18608409 Supplier Spectrum Chemical Mfg Cor Supplier No. HP4124LT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 75-05-8
Molecular Weight (g/mol) 41.05

Specifications

CAS Max % 1
Quantity 4 L
Packaging Amber Glass Bottle
Residue after Evaporation 2 ppm
Titratable Acid 8 μeq/g
Titratable Base 0.6 μeq/g
Water 0.0002
Boiling Point 81.6°C
Percent Purity 0.999
Grade ACS/HPLC
Chemical Name or Material Acetonitrile
Show More Show Less