missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile E CHROMASOLV™, for HPLC, for UV, ≥99.9% (GC), Honeywell Riedel-de Haën™
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Specifications
| Melting Point | -48°C |
| Density | 0.786g/mL at 25°C(lit.) |
| Quantity | 4 x 2.5 L |
| Refractive Index | n20/D 1.344 |
| Linear Formula | CH3CN |
| Vapor Pressure | 72.8mmHg (20°C) |
| Beilstein | 741857 |
| Synonym | methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn |
| Boiling Point | 81°C to 82°C |
| Molecular Weight (g/mol) | 41.053 |
| Show More |
Safety and Handling
| Hazard Statement | P210-P280-P305 + P351 + P338 |
| Precautionary Statement | H225-H302 + H312 + H332-H319 |
| EINECSNumber | 200-835-2 |
| RTECSNumber | AL7700000 |