missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, CHROMASOLV™ (gradient grade +), Solstice

Catalog No. 60048228
Change view
Click to view available options
Quantity:
4 x 4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60048228 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60048228 Supplier Solstice Supplier No. 006834X4L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CHROMASOLV™, gradient grade, suitable for PAH analysis,≥99.9% (GC)

  • Same high quality as CHROMASOLV HPLC
  • Additional testing for HPLC via gradient analysis and UV-VIS spectroscopy
  • Low UV profile by HPLC
  • Low impurity gradient LC baseline

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Absorbance λ215nm : ≤0.01 , λ200nm : ≤4% , λ230nm : ≤0.005
Color Colorless
Density 0.786g/mL at 25°C
Quantity 4 x 4 L
Explosion Limit 0.16
Flash Point 2°C (35.6°F)
Ignition Point 973°F
Packaging Glass Bottle
Vapor Pressure 72.8mmHg (20°C)
Linear Formula CH3CN
UN Number 1648
Beilstein 741857
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile
Solubility Information Completely souble in water
Molecular Weight (g/mol) 41.05
Formula Weight 41.05g/mol
Percent Purity ≥99.9% (GC)
Grade CHROMASOLV™ Gradient +
Chemical Name or Material Acetonitrile, Suitable for PAH analysis
Show More Show Less
Hazard Statement P210-P280-P305 + P351 + P338
Precautionary Statement H225-H302 + H312 + H332-H319
EINECSNumber 200-835-2
RTECSNumber AL7700000