missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, CHROMASOLV™, ≥99.9%, for LC-MS, Honeywell Riedel-de Haën™
CHROMASOLV™ LC-MS
Supplier: Honeywell-Chromasolv 349674X25L
Description
- For LC-MS and LC-UV trace metals analysis
- HPLC / UV-VIS quality control
- Low gradient baseline drift
- Low levels of trace metals
- No non-volatile impurities
Specifications
| Acetonitrile | |
| Colorless | |
| 4 x 2.5 L | |
| 2°C (35.6°F) | |
| C2H3N | |
| MFCD00001878 | |
| UN1648 | |
| methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
| CC#N | |
| 41.053 | |
| CHEBI:38472 | |
| ≥99.9% |
| 75-05-8 | |
| 0.786g/mL at 25°C | |
| 0.16 | |
| 973°F | |
| CH3CN | |
| 72.8 mmHg (20°C) | |
| 741857 | |
| WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
| acetonitrile | |
| 6342 | |
| 41.05g/mol | |
| LC-MS CHROMASOLV |
Chemical Identifiers
| 75-05-8 | |
| 41.053 | |
| WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
| 6342 | |
| acetonitrile |
| C2H3N | |
| MFCD00001878 | |
| methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
| CHEBI:38472 | |
| CC#N |
Safety and Handling
P210-P280-P305 + P351 + P338
RTECSNumber : AL7700000