missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, CHROMASOLV™, ≥99.9%, for LC-MS, Honeywell Riedel-de Haën™

Catalog No. 6000251
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000251 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6000251 Supplier Honeywell Inc Supplier No. 349674X4L
Only null left

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Color Colorless
Density 0.786g/mL at 25°C
Quantity 4 x 4 L
Explosion Limit 0.16
Flash Point 2°C (35.6°F)
Ignition Point 973°F
Linear Formula CH3CN
Vapor Pressure 72.8 mmHg (20°C)
UN Number UN1648
Beilstein 741857
Formula Weight 41.05g/mol
Percent Purity ≥99.9%
Grade LC-MS
Chemical Name or Material Acetonitrile
Show More Show Less
Hazard Statement P210-P280-P305 + P351 + P338
RTECSNumber AL7700000