missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile (Certified ACS), Fisher Chemical™

Catalog No. p-215705
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Glass Bottle
Steel Drum
Steel Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A211 1 L Glass Bottle
A214 4 L Glass Bottle
A2120 20 L Steel Pail
A21200 200 L Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A211 Supplier Thermo Fisher Scientific Supplier No. FLA211
Only null left

Meet or exceeds the latest ACS specifications. Actual lot of analysis on label

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46°C
Color Colorless
Decomposition Information >500°C
Density 0.781g/cm³
Quantity 1 L
Flash Point 12.8°C (55°F)
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.005% max.
Assay Percent Range ≥99.5 %
Titratable Acid 0.008mEq/g max.
Titratable Base 0.0006mEq/g max.
Vapor Pressure 97mbar at 20°C
Linear Formula CH3CN
Viscosity 0.36 cP at 20°C
UN Number 1648
Water 0.3% max.
Boiling Point 81.6°C
Formula Weight 41.05
Percent Purity ≥99.5%
Grade ACS
Chemical Name or Material Acetonitrile
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Irritating to eyes. May cause skin and respiratory tract irritation. May cause central nervous system effects. Liver and kidney injuries may occur. Use personal protective equipment. Use only under a chemical fume hood. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid