missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, B&J Brand™, for HPLC, gradient grade, for spectrophotometry, >99.9%, Honeywell Burdick & Jackson

Catalog No. p-7112021
Change view
Click to view available options
Quantity:
56 L
200 L
Packaging:
Returnable Steel Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000273 56 L Returnable Steel Drum
6000267 200 L Returnable Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000273 Supplier Honeywell Chemicals Supplier No. CS01556
Only null left

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Absorbance 0.05 AU max. at 200 nm, 0.04 AU max. at 210 nm, 0.01 AU max. at 225 nm, 0.005 AU max. at 250 nm, 0.005 AU max. at 350 nm
CAS Max % 100.0000%
Density 0.7822 g/cm3
Boiling Point 82°C
Quantity 56 L
Flash Point 6°C
Packaging Returnable Steel Drum
UN Number UN1648
Beilstein 741857
Water 0.005% max.
Solubility Information Miscible in Water at 20°C
Formula Weight 41
Percent Purity >99.9%
Grade HPLC Gradient
Chemical Name or Material Acetonitrile, B&J Brand™, for HPLC, gradient grade, for spectrophotometry, >99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile
EINECSNumber 200-835-2