missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, for HPLC, gradient grade, for spectrophotometry, >99.9%, Solstice
Specifications
Specifications
| Absorbance | 0.04 AU max. at 210 nm, 0.01 AU max. at 225 nm, 0.005 AU max. at 250 nm, 0.005 AU max. at 350 nm |
| CAS Max % | 100.0000% |
| Density | 0.7822 g/cm3 |
| Quantity | 56 L |
| Flash Point | 6°C |
| Packaging | Returnable Steel Drum |
| Water | 0.005% max. |
| UN Number | UN1648 |
| Beilstein | 741857 |
| Boiling Point | 82°C |
| Show More |
Safety and Handling
| ShelfLife | 547 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetonitrile |
| EINECSNumber | 200-835-2 |